Anti-Oval Cell Marker Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: OC2-3C5]
NOVUNBP1-18968APC
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-3C5
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse