Anti-HSPA5 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-06274R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Glucose-regulated protein 78kDa
- Antigen symbol:GRP78
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3309
- Antigen synonyms:GRP78|BIP|MIF2|HEL-S-89n
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: