Anti-SLC2A9 Rabbit Polyclonal Antibody
Type: Primary
Antigen: SLC2A9 (solute carrier family 2 (facilitated glucose transporter), member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
- Catalog No:
- NOVUNBP1-06271
- NOVUNBP1-06271SS
- Antigen symbol:
- GLUT9
- GLUT9
- Antigen name:
- Glucose Transporter 9
- Glucose Transporter 9
- Antigen synonyms:
- GLUTX|UAQTL2|GLUT9|URATv1
- GLUTX|UAQTL2|GLUT9|URATv1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 56606
- 56606
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together