Anti-AGR2 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNBP1-05933C
- Antibody type:Primary
- Antigen name:Anterior Gradient 2
- Antigen symbol:AGR2
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Gene ID:10551
- Antigen synonyms:AG2|hAG-2|AG-2|Anterior gradient protein 2 homolog|HPC8|Secreted cement gland protein XAG-2 homolog
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: AGR2 (anterior gradient 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse