Anti-TetR Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: TetR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Specifications
- Catalog No:
- NOVUNB600-234SS
- NOVUNB600-234
- Antigen symbol:
- TetR
- TetR
- Antigen name:
- Tet Repressor Protein
- Tet Repressor Protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Multiple
- Multiple
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4924774
- 4924774
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together