Anti-TRAF5 Mouse Monoclonal Antibody [clone: 55A219]
Catalog # NOVUNB600-1107
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 5
- Antigen symbol:TRAF5
- Clonality:Monoclonal
- Clone:55A219
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7188
- Antigen synonyms:RNF84|MGC:39780
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TRAF5 (TNF receptor-associated factor 5)
Clonality: monoclonal
Clone: 55A219
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human