Order Entry
United Kingdom
ContactUsLinkComponent
Anti-BECN1 Rabbit Polyclonal Antibody (DyLight® 488)
Anti-BECN1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB500-249G
Supplier:  Novus Biologicals
Anti-BECN1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB500-249G
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Beclin 1
  • Antigen symbol:
    BECN1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    8678
  • Antigen synonyms:
    Coiled-coil myosin-like BCL2-interacting protein|APG 6|VPS30|Beclin1 autophagy|ATG6|CTIP-1 COUP-TF interacting protein 1|Protein GT197
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat