Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]
 
undefined
Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]

 

  • Catalog No:
  • NOVUNB400-146
  • NOVUNB400-146SS
  • Antigen symbol:
  • THOP1
  • THOP1
  • Antigen name:
  • Thimet Oligopeptidase 1
  • Thimet Oligopeptidase 1
  • Antigen synonyms:
  • MP78|TOP|MEPD_HUMAN|EP24.15
  • MP78|TOP|MEPD_HUMAN|EP24.15
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 4D6
  • 4D6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 7064
  • 7064
  • Isotype:
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together