Anti-RNA polymerase II Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4H8]
NOVUNB200-598PE
- Antibody type:Primary
- Antigen name:Polymerase (RNA) II (DNA directed) polypeptide A, 220kDa
- Antigen symbol:POLR2A
- Clonality:Monoclonal
- Clone:4H8
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Gene ID:5430
- Antigen synonyms:hsRPB1|RPOL2|POLRA|RPBh1|RpIILS|RPO2|RPB1|POLR2|hRPB220
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: RNA polymerase II
Clonality: monoclonal
Clone: 4H8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: