Anti-TRIM29 Rabbit Polyclonal Antibody
Catalog # NOVUNB120-22207
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 29
- Antigen symbol:TRIM29
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Gene ID:23650
- Antigen synonyms:ATDC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TRIM29 (tripartite motif containing 29)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human