Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 1B20]
NOVUNB110-89062IR
- Antibody type:Primary
- Antigen name:Secreted Phosphoprotein 1
- Antigen symbol:SPP1
- Clonality:Monoclonal
- Clone:1B20
- Conjugation:DyLight® 755
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Gene ID:6696
- Antigen synonyms:ETA-1|OPN|BNSP|BSPI
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: monoclonal
Clone: 1B20
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: