Anti-SPP1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 1B20]
Catalog # NOVUNB110-89062C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Secreted Phosphoprotein 1
- Antigen symbol:SPP1
- Clonality:Monoclonal
- Clone:1B20
- Conjugation:DyLight® 650
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Gene ID:6696
- Antigen synonyms:ETA-1|Opnl|Bsp|OP|2AR|BNSP|BSPI|Eta|Ric|Apl-1|Opn|Spp-1|Osteopontin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: monoclonal
Clone: 1B20
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: