Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 405) [clone: FDO66Q]
Catalog # NOVUNB110-78644V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
- Antigen symbol:HSD3B1
- Clonality:Monoclonal
- Clone:FDO66Q
- Conjugation:DyLight® 405
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Gene ID:3283
- Antigen synonyms:HSDB3A|3BETAHSD|HSD3B|HSDB3|SDR11E1|I
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone: FDO66Q
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: