Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody (DyLight® 350) [clone: SE-1]
NOVUNB110-68095UV
- Antibody type:Primary
- Antigen name:Hepatic Sinusoidal Endothelial Cells
- Clonality:Monoclonal
- Clone:SE-1
- Conjugation:DyLight® 350
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: Hepatic Sinusoidal Endothelial Cells
Clonality: monoclonal
Clone: SE-1
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse, Rat