Anti-SLC9A8 Mouse Monoclonal Antibody (DyLight® 405) [clone: 7A11]
Catalog # NOVUNB110-62091V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8
- Antigen symbol:Solute carrier family 9, member 8
- Clonality:Monoclonal
- Clone:7A11
- Conjugation:DyLight® 405
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:23315
- Antigen synonyms:NHE-8|NHE8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC9A8 (solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8)
Clonality: monoclonal
Clone: 7A11
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat