Anti-APOA5 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 4H8H8E2]
NOVUNB110-57307F
- Antibody type:Primary
- Antigen name:Apolipoprotein A5
- Antigen symbol:APOA5
- Clonality:Monoclonal
- Clone:4H8H8E2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:116519
- Antigen synonyms:RAP3|APOAV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: monoclonal
Clone: 4H8H8E2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human