Anti-LRRK2 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB110-55289V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:leucine-rich repeat kinase 2
- Antigen symbol:LRRK2
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:120892
- Antigen synonyms:DARDARIN|PARK8|ROCO2|RIPK7|AURA17
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse