Anti-rRNA Mouse Monoclonal Antibody (DyLight® 755) [clone: Y10b]
NOVUNB100-662IR
- Antibody type:Primary
- Antigen symbol:rRNA
- Clonality:Monoclonal
- Clone:Y10b
- Conjugation:DyLight® 755
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3 kappa
- Reactivity:Multiple
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: rRNA
Clonality: monoclonal
Clone: Y10b
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Pan