Anti-Survivin Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56167
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Baculoviral IAP Repeat Containing 5
- Antigen symbol:BIRC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:332
- Antigen synonyms:Apoptosis Inhibitor 4|SVV|TIAP|IAP4|API4|Baculoviral IAP repeat containing 5|EPR 1|Apoptosis inhibitor survivin|BIRC 5
- Storage buffer:Whole antisera
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
The Survivin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Survivin. This antibody reacts with human, mouse, rat, gerbil. The Survivin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000
Type: Primary
Antigen: Survivin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Gerbil