Anti-cIAP-2/HIAP-1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56132
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 3
- Antigen symbol:cIAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:330
- Antigen synonyms:MIHC|MALT2|HAIP1|API2|AIP1|RNF49|CIAP2|HIAP1|c-IAP2
- Storage buffer:Neat whole antisera
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
The cIAP-2 / HIAP-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to cIAP-2/HIAP-1. This antibody reacts with human. The cIAP-2/HIAP-1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000
Type: Primary
Antigen: cIAP-2/HIAP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human