Anti-SIRT6 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB100-2523V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sirtuin 6
- Antigen symbol:SIRT6
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:51548
- Antigen synonyms:SIR2L6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human