Anti-DSG3 Mouse Monoclonal Antibody [clone: 5G11]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-1643SS
- NOVUNB100-1643
- Antigen symbol:
- DSG3
- DSG3
- Antigen name:
- Desmoglein 3
- Desmoglein 3
- Antigen synonyms:
- 130 kDa pemphigus vulgaris antigen|130-kD pemphigus vulgaris antigen|CDHF6|Cadherin family member 6|desmoglein-3|PVA|pemphigus vulgaris antigen
- 130 kDa pemphigus vulgaris antigen|130-kD pemphigus vulgaris antigen|CDHF6|Cadherin family member 6|desmoglein-3|PVA|pemphigus vulgaris antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 5G11
- 5G11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1830
- 1830
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together