Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 650) [clone: H1alpha67]
Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 650) [clone: H1alpha67]
  NOVUNB100-105C
undefined
Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 650) [clone: H1alpha67]
  NOVUNB100-105C

 

  • Antibody type:
    Primary
  • Antigen name:
    HIF1 alpha
  • Antigen symbol:
    HIF1A
  • Clonality:
    Monoclonal
  • Clone:
    H1alpha67
  • Conjugation:
    DyLight® 650
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    3091
  • Antigen synonyms:
    PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: