Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ATM Rabbit Polyclonal Antibody (DyLight® 550)
Anti-ATM Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB100-104R
Supplier:  Novus Biologicals
Anti-ATM Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB100-104R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ataxia Telangiectasia mutated
  • Antigen symbol:
    ATM
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    472
  • Antigen synonyms:
    ATM Protein Kinase pS1981
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µL
  • Pk:
    0,05 mL

Specifications

About this item

Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse