Anti-Laminin 2 alpha Rabbit Polyclonal Antibody
Catalog # BSBTPA2160
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Laminin 2 alpha
- Antigen symbol:LAMA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:152 - 170
- Sequence:WILERSLDDVEYKPWQYHA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Laminin 2 alpha (152-170aa WILERSLDDVEYKPWQYHA), identical to the related mouse sequence, and different from the related rat sequence by one amino acid
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Laminin subunit alpha-2(LAMA2) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Laminin 2 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat