Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-BrdU Mouse Monoclonal Antibody [clone: Bu20a]
  ABCAAB136650-100
 :  Abcam
undefined
Anti-BrdU Mouse Monoclonal Antibody [clone: Bu20a]
  ABCAAB136650-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Bromodeoxyuridine
  • Antigen symbol:
    BrdU
  • Clonality:
    Monoclonal
  • Clone:
    Bu20a
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Mouse monoclonal [Bu20a] to BrdU.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone: Bu20a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: