Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-NMDAR2A (phospho Y1325) Rabbit Polyclonal Antibody
Catalog # ABCAAB106590-100
Supplier:  Abcam
undefined
Anti-NMDAR2A (phospho Y1325) Rabbit Polyclonal Antibody
Catalog # ABCAAB106590-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2A
  • Antigen symbol:
    GRIN2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal to NMDAR2A (phospho Y1325).

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GRIN2A (glutamate receptor, ionotropic, N-methyl D-aspartate 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat