Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SIGLEC8 Mouse Monoclonal Antibody [clone: 7C9]
Anti-SIGLEC8 Mouse Monoclonal Antibody [clone: 7C9]
Catalog # ABCAAB103398-50
Supplier:  Abcam
undefined
Anti-SIGLEC8 Mouse Monoclonal Antibody [clone: 7C9]
Catalog # ABCAAB103398-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sialic acid binding Ig-like lectin 8
  • Antigen symbol:
    SIGLEC8
  • Clonality:
    Monoclonal
  • Clone:
    7C9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal [7C9] to SIGLEC8.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SIGLEC8
Clonality: Monoclonal
Clone: 7C9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human