Anti-Histone H3.3 Rabbit monoclonal antibody unconjugated [clone: EPR1873]
Catalog # ABCAAB92628-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Histone H3.3
- Clonality:Monoclonal
- Clone:EPR1873
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- ChIP:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3.3
Clonality: monoclonal
Clone: EPR1873
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: