Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Glycogen synthase 1 Rabbit monoclonal antibody unconjugated [clone: EP852Y]
Anti-Glycogen synthase 1 Rabbit monoclonal antibody unconjugated [clone: EP852Y]
Catalog # ABCAAB81230-100
Supplier:  Abcam
Anti-Glycogen synthase 1 Rabbit monoclonal antibody unconjugated [clone: EP852Y]
Catalog # ABCAAB81230-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glycogen Synthase 1
  • Antigen symbol:
    GYS1
  • Clonality:
    Monoclonal
  • Clone:
    EP852Y
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glycogen synthase 1
Clonality: monoclonal
Clone: EP852Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: