Order Entry
United Kingdom
ContactUsLinkComponent
Anti-GFP Rabbit polyclonal antibody Sepharose
Anti-GFP Rabbit polyclonal antibody Sepharose
  ABCAAB69314-25
 :  Abcam
Anti-GFP Rabbit polyclonal antibody Sepharose
  ABCAAB69314-25
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Green Fluorescence Protein
  • Antigen symbol:
    GFP
  • Clonality:
    Polyclonal
  • Conjugation:
    Sepharose
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    25 µg
  • Pk:
    25 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: Sepharose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: