Anti-Histone H2B (acetyl K12 + K12 + K16 + K23 + K23 + K23 + K23 + K23 + K5 + K7) Rabbit Monoclonal Antibody [clone: EP819Y]
Catalog # ABCAAB52985-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:histone cluster 2, H2be
- Antigen symbol:HIST2H2BE
- Clonality:Monoclonal
- Clone:EP819Y
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [EP819Y] to Histone H2B - ChIP Grade.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: EP819Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human