Order Entry
United Kingdom
ContactUsLinkComponent
Anti-cGMP Sheep Polyclonal Antibody
Anti-cGMP Sheep Polyclonal Antibody
  ABCAAB19226-2
 :  Abcam
Anti-cGMP Sheep Polyclonal Antibody
  ABCAAB19226-2
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic guanosine monophosphate
  • Antigen symbol:
    cGMP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    2 ml
  • Pk:
    2 mL

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: cGMP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: