Anti-Kappa light chain Mouse Monoclonal Antibody [clone: 4G7]
ABCAAB1936-500
: Abcam
- Antibody type:Primary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Monoclonal
- Clone:4G7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Kappa light chain
Clonality: monoclonal
Clone: 4G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human