Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Nuclear Matrix Protein p84 Mouse Monoclonal Antibody [clone: 5E10]
Anti-Nuclear Matrix Protein p84 Mouse Monoclonal Antibody [clone: 5E10]
Catalog # ABCAAB487-100
Supplier:  Abcam
Anti-Nuclear Matrix Protein p84 Mouse Monoclonal Antibody [clone: 5E10]
Catalog # ABCAAB487-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear Matrix Protein p84
  • Clonality:
    Monoclonal
  • Clone:
    5E10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Nuclear Matrix Protein p84
Clonality: monoclonal
Clone: 50000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse