Anti-ANP32E Rabbit Polyclonal Antibody
Catalog # BIRBORB86677-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Acidic (leucine-rich) nuclear phosphoprotein 32 family, member E
- Antigen symbol:ANP32E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 2.5 ?g/ml; Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: ANP32E (acidic (leucine-rich) nuclear phosphoprotein 32 family, member E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat