Anti-ABCC4 Goat Polyclonal Antibody
BIRBORB86394-50
: Biorbyt
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 4
- Antigen symbol:ABCC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide C-NGQPSTLTIFETAL from the C-terminus of human ABCC4/MRP4
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1:64000; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: ABCC4 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat