Anti-GPR82 Rabbit Polyclonal Antibody
BIRBORB79865-50
: Biorbyt
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 82
- Antigen symbol:GPR82
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 15 amino acid peptide from 2nd cytoplasmic domain of human GPR82
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 6-16 ?g/ml
Type: Primary
Antigen: GPR82 (G protein-coupled receptor 82)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human