Anti-HSD3B7 Rabbit Polyclonal Antibody
Catalog # BIRBORB77927-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
- Antigen symbol:HSD3B7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant protein
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:20000-1:80000; Immunochemistry: 1:100-1:500
Type: Primary
Antigen: HSD3B7 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human