Anti-Complement C4d Mouse Monoclonal Antibody
Catalog # BIRBORB76603-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Complement C4d
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:Purified antibody containing PBS, 10mM sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Complement C4d for normal human plasma
- Purification:purified by Protein G chromatography
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Complement C4d
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human