Anti-SOCS1 Rabbit Polyclonal Antibody
Catalog # BIRBORB74706-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Suppressor of Cytokine Signaling 1
- Antigen symbol:SOCS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/mL
- Shipping temperature:Ice
- Immunogen:Raised against a 17 amino acid peptide from near the amino terminus of human SOCS1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human