Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SCNN1A Rabbit Polyclonal Antibody
Anti-SCNN1A Rabbit Polyclonal Antibody
Catalog # BIRBORB74006-100
Supplier:  Biorbyt
undefined
Anti-SCNN1A Rabbit Polyclonal Antibody
Catalog # BIRBORB74006-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    sodium channel, non-voltage-gated 1 alpha subunit
  • Antigen symbol:
    SCNN1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: SCNN1A (sodium channel, non-voltage-gated 1 alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human