Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ODC1 Rabbit Polyclonal Antibody
Anti-ODC1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-ODC1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB136017-100
  • BIRBORB136017-50
  • BIRBORB136017-200
  • Antigen symbol:
  • ODC1
  • ODC1
  • ODC1
  • Antigen name:
  • ornithine decarboxylase 1
  • ornithine decarboxylase 1
  • ornithine decarboxylase 1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together