Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SLC7A11 Rabbit Polyclonal Antibody (RBITC)
Anti-SLC7A11 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131672-100
Supplier:  Biorbyt
Anti-SLC7A11 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131672-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11
  • Antigen symbol:
    Solute carrier family 7, member 11
  • Clonality:
    Polyclonal
  • Conjugation:
    RBITC (Rhodamine B Isothiocyanate)
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    52 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: