Anti-OLR1 Rabbit Polyclonal Antibody (RBITC)
BIRBORB131494-100
: Biorbyt
- Antibody type:Primary
- Antigen name:oxidized low density lipoprotein (lectin-like) receptor 1
- Antigen symbol:OLR1
- Clonality:Polyclonal
- Conjugation:RBITC (Rhodamine B Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:31/50 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human LOX-1 C-terminus.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: OLR1 (oxidized low density lipoprotein (lectin-like) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: