Anti-HMGCR Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
BIRBORB130924-100
: Biorbyt
- Antibody type:Primary
- Antigen name:3-hydroxy-3-methylglutaryl-CoA reductase
- Antigen symbol:HMGCR
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy7®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Modification:phospho-Ser872
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:97 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human HMGCR around the phosphorylation site of Ser872.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: HMGCR (3-hydroxy-3-methylglutaryl-CoA reductase)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat