Anti-LPAR2 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
BIRBORB130861-100
: Biorbyt
- Antibody type:Primary
- Antigen name:lysophosphatidic acid receptor 2
- Antigen symbol:LPAR2
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy7®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:39 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human EDG4/LPA2.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: LPAR2 (lysophosphatidic acid receptor 2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: