Anti-BrdU Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # BIRBORB113495-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: