Anti-Kelch-like 1 protein Rabbit Polyclonal Antibody
Catalog # BIRBORB108967-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Kelch-like 1 Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Rabbit polyclonal KLHL1 antibody was raised against a 14 amino acid peptide from near the amino-terminus of human KLHL1.
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Kelch-like 1 protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: