Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HA tag Mouse Monoclonal Antibody
Anti-HA tag Mouse Monoclonal Antibody
Supplier:  Biorbyt
Certificates
Anti-HA tag Mouse Monoclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22866-0.5
  • BIRBORB22866-100
  • BIRBORB22866-0.2
  • Antigen symbol:
  • HA
  • HA
  • HA
  • Antigen name:
  • HA tag
  • HA tag
  • HA tag
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (YPYDVPDYA) coupled to KLH.
  • Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (YPYDVPDYA) coupled to KLH.
  • Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (YPYDVPDYA) coupled to KLH.
  • Sequence:
  • YPYDVPDYA
  • YPYDVPDYA
  • YPYDVPDYA
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Frequently Bought Together